Dimethyl sulphoxide-[D₆], Sigma-Aldrich®
Supplier: Merck
Synonyms: Dimethyl sulphoxide-[²H₆], Hexadeuterodimethyl sulphoxide, DMSO-D6, DMSO-d6, Dimethyl sulphoxide-d6
M-Clarity™ quality level = MQ100
185965-10G 185965-25G 185965-50G
SIAL185965-10GEA 50.4 EUR
SIAL185965-10G SIAL185965-25G SIAL185965-50G
Dimethyl sulphoxide-[D₆], Sigma-Aldrich®
Dimethyl sulphoxide-[D6]
Formula: D₃CS(O)CD₃
Boiling Pt: 189 °C (5 mmHg)
Melting Pt: 20.2 °C
Density: 1.19 g/cm³ (20 °C)
Flash Pt: 82 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002090
CAS Number: 2206-27-1
EINECS: 218-617-0