Dichloromethane-[D₂], Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Dichloromethane-d2, Methylene chloride-D2, Dichloromethane-[²H₂], Dideuteromethylene chloride, Methylene chloride-d2
MQ200
177865-10G 177865-25G
SIAL177865-10GEA 208 EUR
SIAL177865-10G SIAL177865-25G
Dichloromethane-[D₂], Sigma-Aldrich®
Dichloromethane-[D2]
Formula: CD₂Cl₂
MW: 86.92 g/mol
Boiling Pt: 40
Melting Pt: –97 °C
Density: 1.36 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000882
CAS Number: 1665-00-5
EINECS: 216-776-0
UN: 1593
ADR: 6.1 III