Iodoethane[D5], Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
324582-1G
324582-5G
SIAL324582-1GEA
130
EUR
SIAL324582-1G
SIAL324582-5G
Iodoethane[D5], Sigma-Aldrich®
Iodoethane-[D5]
Formula:
D₃CCD₂I Boiling Pt: 69 °C (1013 hPa) Melting Pt: –108 °C Density: 2.012 g/cm³ (20 °C) |
CAS Number:
6485-58-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...