Bromobenzene-[D₅], Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

175730-25G 175730-5G
SIAL175730-25GEA 2380 EUR
SIAL175730-25G SIAL175730-5G
Bromobenzene-[D₅], Sigma-Aldrich®
Bromobenzene-[D5]
Formula: C₆D₅Br
Boiling Pt: 156.6 °C (1013 hPa)
Melting Pt: –31 °C
Density: 1.55 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000056
CAS Number: 4165-57-5
EINECS: 224-013-8
UN: 2514
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR