1,3,5-Trimethylbenzene-[D12], Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

372374-1G 372374-5G
SIAL372374-1GEA 227 EUR
SIAL372374-1G SIAL372374-5G
1,3,5-Trimethylbenzene-[D12], Sigma-Aldrich®
1,3,5-Trimethylbenzene-[D12]
Formula: C₆D₃(CD₃)₃
CAS Number: 69441-16-3
UN: 2325
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR