Chloropentacarbonylrhenium, Sigma-Aldrich®

Supplier: Merck

Synonyms: Pentacarbonylchlororhenium(I), Chloropentacarbonylrhenium(I), Rhenium pentacarbonyl chloride

M-Clarity™ quality level = MQ100

327611-1G 327611-5G
SIAL327611-1GEA 153 EUR
SIAL327611-1G SIAL327611-5G
Chloropentacarbonylrhenium, Sigma-Aldrich®
Chloropentacarbonylrhenium
Formula: C₅ClO₅Re
MDL Number: MFCD00013296
CAS Number: 14099-01-5
EINECS: 237-948-1
UN: 3466
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR