D-(+)-Maltose monohydrate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Maltobiose hydrate, Malt Sugar, beta-Maltose monohydrate, Maltose monohydrate, (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal hydrate, 4-O-α-D-Glucopyranosyl-D-glucose hydrate
M-Clarity™ quality level = MQ200
M5895-1KG M5895-500G
SIALM5895-1KGEA 378 EUR
SIALM5895-1KG SIALM5895-500G
D-(+)-Maltose monohydrate, Sigma-Aldrich®
D(+)-Maltose monohydrate
Formula: C₁₂H₂₂O₁₁·1H₂O
Melting Pt: 160…165 °C
Storage Temperature: Ambient
MDL Number: MFCD00149343
CAS Number: 6363-53-7
EINECS: 200-716-5
Merck Index: 13,05736
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f861d7adf53134a0c485b5942d3be34c4d0b571&vid=16e5c4425a7a8e158a4200470a45483c7ff5781e&prid=web&clid=SAPDAM