D-(+)-Trehalose dihydrate, Millipore®
Supplier: Merck
Synonyms: Mycose, α,α-Trehalose, 1-O-α-D-Glucopyranosyl-α-D-glucopyranoside, α-D-Glucopyranosyl-α-D-glucopyranoside, (2R,2'R,3S,3'S,4S,4'S,5R,5'R,6R,6'R)-6,6'-Oxybis(2-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol) dihydrate
M-Clarity™ quality level = MQ200
90210-250G 90210-50G
SIAL90210-250GEA 811 EUR
SIAL90210-250G SIAL90210-50G
D-(+)-Trehalose dihydrate, Millipore®
D(+)-Trehalose dihydrate
Formula: C₁₂H₂₂O₁₁·2H₂O
Melting Pt: 97…99 °C
Storage Temperature: Ambient
MDL Number: MFCD00071594
CAS Number: 6138-23-4
EINECS: 202-739-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af741ee934bfe7c82395d31c21e2c19f0738d0f0&vid=59183c854dcfa367e15d46861ef4b9a7028540a9&prid=web&clid=SAPDAM