D-(+)-Sucrose, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ300
S1888-1KG
S1888-500G
S1888-5KG
SIALS1888-1KGEA
188
EUR
SIALS1888-1KG
SIALS1888-500G
SIALS1888-5KG
D-(+)-Sucrose, Sigma-Aldrich®
D(+)-Sucrose
Formula:
C₁₂H₂₂O₁₁ Melting Pt: 169…170 °C Density: 1.29 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00006626 CAS Number: 57-50-1 EINECS: 200-334-9 Merck Index: 13,08966 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...