D-(+)-Sucrose, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ300
S5016-1KG
S5016-2.5KG
S5016-25G
S5016-4X2.5KG
S5016-500G
S5016-6X500G
SIALS5016-1KGEA
160
EUR
SIALS5016-1KG
SIALS5016-2.5KG
SIALS5016-25G
SIALS5016-4X2.5KG
SIALS5016-500G
SIALS5016-6X500G
D-(+)-Sucrose, Sigma-Aldrich®
D(+)-Sucrose
Formula:
C₁₂H₂₂O₁₁ Melting Pt: 169…170 °C Density: 1.29 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00006626 CAS Number: 57-50-1 EINECS: 200-334-9 Merck Index: 13,08966 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...