D-(+)-Sucrose, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ300
S7903-250G S7903-1KG S7903-5KG
SIALS7903-250GEA 46.9 EUR
SIALS7903-250G SIALS7903-1KG SIALS7903-5KG
D-(+)-Sucrose, Sigma-Aldrich®
D(+)-Sucrose
Formula: C₁₂H₂₂O₁₁
Melting Pt: 169…170 °C
Density: 1.29 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006626
CAS Number: 57-50-1
EINECS: 200-334-9
Merck Index: 13,08966
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=971b1a9d1f02c06efd63ac665dfb191799fbd5ac&vid=2ddc1affd401bacf02244ae0cb620ff18e440965&prid=web&clid=SAPDAM