D-(+)-Sucrose, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
S9378-10KG
S9378-1KG
S9378-500G
S9378-5KG
S9378-10MG
SIALS9378-10KGEA
612
EUR
SIALS9378-10KG
SIALS9378-1KG
SIALS9378-500G
SIALS9378-5KG
SIALS9378-10MG
D-(+)-Sucrose, Sigma-Aldrich®
D(+)-Sucrose
Formula:
C₁₂H₂₂O₁₁ Melting Pt: 169…170 °C Density: 1.29 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00006626 CAS Number: 57-50-1 EINECS: 200-334-9 Merck Index: 13,08966 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...