D-(+)-Galactose, Sigma-Aldrich®
Supplier: Merck
Synonyms: D-Galactose, (2R,3S,4S,5R)-2,3,4,5,6-Pentahydroxyhexanal, (2R,3S,4S,5R)-Pentahydroxyhexanal
M-Clarity™ quality level = MQ300
G0625-100G G0625-1KG G0625-500G G0625-5KG
SIALG0625-100GEA 72.3 EUR
SIALG0625-100G SIALG0625-1KG SIALG0625-500G SIALG0625-5KG
D-(+)-Galactose, Sigma-Aldrich®
D(+)-Galactose
Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Boiling Pt: 527
Melting Pt: 163…169 °C
Density: 1.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00151230
CAS Number: 59-23-4
EINECS: 200-416-4
Merck Index: 13,04356
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=508b1d21a6801ddfb1a5f13a64cb06e6f95e8922&vid=3c02003b1d12c9bfd11c2e6faf5cc37d4325d208&prid=web&clid=SAPDAM