Starch, hydrolysed acc. Smithies

Supplier: Roth Carl

9480.1 9480.2 9480.4
ROTH9480.1EA 0 EUR
ROTH9480.1 ROTH9480.2 ROTH9480.4
Starch, hydrolysed acc. Smithies
Starch
Formula: (C₆H₁₀O₅)n
MW: 162.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00082026
CAS Number: 9005-25-8
EINECS: 232-679-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR