Starch Potato
Supplier: MP Biomedicals
02102954.5 0210295402 0210295405
ICNA02102954.5EA 37.4 EUR
ICNA02102954.5 ICNA0210295402 ICNA0210295405
Starch Potato
Starch
Formula: (C₆H₁₀O₅)n
MW: 162.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00082026
CAS Number: 9005-25-8
EINECS: 232-679-6