Starch (from potato)
Supplier: Roth Carl
9441.1 9441.2
ROTH9441.1EA 24.7 EUR
ROTH9441.1 ROTH9441.2
Starch (from potato)
Starch
Formula: (C₆H₁₀O₅)n
MW: 162.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00082026
CAS Number: 9005-25-8
EINECS: 232-679-6