Starch (from potato), Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
S4251-2KG
S4251-5KG
SIALS4251-2KGEA
126
EUR
SIALS4251-2KG
SIALS4251-5KG
Starch (from potato), Sigma-Aldrich®
Starch
Formula:
(C₆H₁₀O₅)n Storage Temperature: Ambient |
MDL Number:
MFCD00082026 CAS Number: 9005-25-8 EINECS: 232-679-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...