Starch (from corn), Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

S4180-100G S4180-1KG S4180-500G
SIALS4180-100GEA 41.7 EUR
SIALS4180-100G SIALS4180-1KG SIALS4180-500G
Starch (from corn), Sigma-Aldrich®
Starch
Formula: (C₆H₁₀O₅)n
MW: 162.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00082026
CAS Number: 9005-25-8
EINECS: 232-679-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR