Starch Corn, Sigma-Aldrich®
Supplier: Merck
MQ200
S9679-1KG S9679-250G
SIALS9679-1KGEA 262 EUR
SIALS9679-1KG SIALS9679-250G
Starch Corn, Sigma-Aldrich®
Starch
Formula: (C₆H₁₀O₅)n
MW: 162.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00082026
CAS Number: 9005-25-8
EINECS: 232-679-6