Bovine serum albumin (BSA)

Supplier: Merck

B4287-25G B4287-5G
SIALB4287-25GEA 200 EUR
SIALB4287-25G SIALB4287-5G
Bovine serum albumin (BSA)
Proteins and Peptides

Order Now

SPECIFICATIONS

Protein/peptide name Bovine Serum Albumin
Environmentally Preferable
Protein synonyms BSA|Fraction V|Cohn Fraction V
Species Bovine
Source Serum
CAS No. 9048-46-8
90604-29-8
Grade for molecular biology
Colour light yellow
Storage conditions Refrigerator

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR