Polydimethylsiloxane, 100000 cSt, Sigma-Aldrich®
Supplier: Merck
Synonyms: Dimethylpolysiloxane, PDMS, Silicone oil, Dimethicone
M-Clarity™ quality level = MQ100
378437-1L 378437-250ML
SIAL378437-1LEA 688 EUR
SIAL378437-1L SIAL378437-250ML
Polydimethylsiloxane, 100000 cSt, Sigma-Aldrich®
Polydimethylsiloxane
Formula: (CH₃)₃SiO(Si(CH₃)₂O)n
MDL Number: MFCD00084411
CAS Number: 63148-62-9