Polydimethylsiloxane, 10000 cSt, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Dimethylpolysiloxane, PDMS, Silicone oil, Dimethicone
M-Clarity™ quality level = MQ100
378402-1L
378402-250ML
SIAL378402-1LEA
358
EUR
SIAL378402-1L
SIAL378402-250ML
Polydimethylsiloxane, 10000 cSt, Sigma-Aldrich®
Polydimethylsiloxane
Formula:
(CH₃)₃SiO(Si(CH₃)₂O)n Boiling Pt: >140 °C (0.002 mmHg)) (lit.) Density: 0.971 g/ml (25 °C) |
MDL Number:
MFCD00132673 CAS Number: 63148-62-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...