Polyethylene glycol 1,000
Supplier: Thermo Fisher Scientific
CAS No.: 25322-68-3
Molecular Formula: H(OCH2CH2)nOH
Flash Point: 260°(500°F)
Physical Form: Powder/low melting solid
Density: 1.101
Appearance: White
Odour: Odourless
pH: 5-7
Melting Point: 35-40°
Merck Reference: 14,7568
MDL No.: MFCD01779605
Formula:
H(OCH₂CH₂)nOH Boiling Pt: ∼250 °C (1013 hPa) Melting Pt: 35…40 °C Density: 1.101 Flash Pt: 260 °C (500 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00081839 CAS Number: 25322-68-3 EINECS: 500-038-2 Merck Index: 14,7568 |