Polyethylene glycol 10,000, Sigma-Aldrich®
Supplier: Merck
Synonyms: PEG 10000, Ethylene glycol polymer 10000, PEG 10,000
92897-1KG-F 92897-250G-F
SIAL92897-1KG-FEA 115 EUR
SIAL92897-1KG-F SIAL92897-250G-F
Polyethylene glycol 10,000, Sigma-Aldrich®
Polyethylene glycol 10,000
Formula: H(OCH₂CH₂)nOH
MW: 10000 g/mol
Melting Pt: 63
Density: 1.2 g/cm³ (20 °C)
Flash Pt: 265
Storage Temperature: Ambient
MDL Number: MFCD00081839
CAS Number: 25322-68-3
EINECS: 500-038-2