Polyethylene glycol 1,450
Supplier: Thermo Fisher Scientific
Synonyms: PEG 1,450, Ethylene glycol polymer 1450, PEG 1450
418040010 418040030 418041000 418045000
ACRO418040010EA 40 EUR
ACRO418040010 ACRO418040030 ACRO418041000 ACRO418045000
Polyethylene glycol 1,450
Polyethylene glycol 1,450
Formula: H(OCH₂CH₂)nOH
MW: 1450 g/mol
Melting Pt: 44…48 °C
Storage Temperature: Ambient
MDL Number: MFCD00081839
CAS Number: 25322-68-3
EINECS: 500-038-2
Order Now
Specification Test Results
Appearance White Waxy flakes
Infrared spectrum Conforms
Water ≤0.5 % (K.F.)
Hydroxyl value 70 to 80 mg KOH/g