Polyethylene glycol 1,500, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: PEG 1500, PEG 1,500
M-Clarity™ quality level = MQ200
81210-1KG 81210-5KG
SIAL81210-1KGEA 106 EUR
SIAL81210-1KG SIAL81210-5KG
Polyethylene glycol 1,500, Sigma-Aldrich®
Polyethylene glycol 1,500
Formula: H(OCH₂CH₂)nOH
MW: 1500 g/mol
Boiling Pt: ∼250 °C (1013 hPa)
Melting Pt: 44…48 °C
Density: 1.13 g/cm³ (20 °C)
Flash Pt: 260
Storage Temperature: Ambient
MDL Number: MFCD00081839
CAS Number: 25322-68-3
EINECS: 500-038-2