Polyethylene glycol 200, Sigma-Aldrich®

Supplier: Merck

Synonyms: PEG 200, Ethylene glycol polymer average Mn 200

88440-1L-F 88440-250ML-F
SIAL88440-1L-FEA 107 EUR
SIAL88440-1L-F SIAL88440-250ML-F
Polyethylene glycol 200, Sigma-Aldrich®
Polyethylene glycol 200
Formula: H(OCH₂CH₂)nOH
MW: 200 g/mol
Boiling Pt: ∼250 °C (1013 hPa)
Melting Pt: –55…–40 °C
Density: 1.124 g/cm³ (20 °C)
Flash Pt: 180
Storage Temperature: Ambient
MDL Number: MFCD01779596
CAS Number: 25322-68-3
EINECS: 500-038-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR