Polyethylene glycol 2,000, Sigma-Aldrich®
Supplier: Merck
Synonyms: PEG 2000, Ethylene glycol polymer 2000, PEG 2,000, Ethylene glycol polymer average Mn 2000
84797-1KG-F 84797-250G-F
SIAL84797-1KG-FEA 121 EUR
SIAL84797-1KG-F SIAL84797-250G-F
Polyethylene glycol 2,000, Sigma-Aldrich®
Polyethylene glycol 2,000
Formula: H(OCH₂CH₂)nOH
Melting Pt: 49…52 °C
Density: 1.21 g/cm³ (20 °C)
Flash Pt: 250 °C
Storage Temperature: Ambient
MDL Number: MFCD00081839
CAS Number: 25322-68-3
EINECS: 500-038-2