Polyethylene glycol 400, Sigma-Aldrich®
Supplier: Merck
Synonyms: PEG 400
M-Clarity™ quality level = MQ200
202398-250G 202398-500G 202398-20KG
SIAL202398-250GEA 28.3 EUR
SIAL202398-250G SIAL202398-500G SIAL202398-20KG
Polyethylene glycol 400, Sigma-Aldrich®
Polyethylene glycol 400
Formula: H(OCH₂CH₂)nOH
MW: 400 g/mol
Boiling Pt: ∼250 °C (1013 hPa)
Melting Pt: 4…8 °C
Density: 1.13 g/cm³ (20 °C)
Flash Pt: 245
Storage Temperature: Ambient
MDL Number: MFCD00081839
CAS Number: 25322-68-3
EINECS: 500-038-2