Polyethylene glycol 600, Sigma-Aldrich®
Supplier: Merck
Synonyms: Ethylene glycol polymer average Mn 600, PEG 600
87333-1KG-F 87333-250G-F
SIAL87333-1KG-FEA 131 EUR
SIAL87333-1KG-F SIAL87333-250G-F
Polyethylene glycol 600, Sigma-Aldrich®
Polyethylene glycol 600
Formula: H(OCH₂CH₂)nOH
MW: 600 g/mol
Boiling Pt: ∼250 °C (1013 hPa)
Melting Pt: 17…22 °C
Density: 1.13 g/cm³ (20 °C)
Flash Pt: 270
Storage Temperature: Ambient
MDL Number: MFCD00081839
CAS Number: 25322-68-3
EINECS: 500-038-2