Polyethylene glycol 6,000, Sigma-Aldrich®
Supplier: Merck
Synonyms: PEG 6,000, PEG 6000
M-Clarity™ quality level = MQ200
81255-1KG 81255-2.5KG 81255-250G
SIAL81255-1KGEA 142 EUR
SIAL81255-1KG SIAL81255-2.5KG SIAL81255-250G
Polyethylene glycol 6,000, Sigma-Aldrich®
Polyethylene glycol 6,000
Formula: H(OCH₂CH₂)nOH
MW: 6000 g/mol
Boiling Pt: ∼250 °C (1013 hPa)
Melting Pt: 55…62 °C
Density: 1.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD01779614
CAS Number: 25322-68-3
EINECS: 500-038-2