TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether), Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Polyethyleneglycol-[4-(1,1,3,3-tetramethylbutyl)phenyl]-ether, (p-tert-Octylphenoxy)polyethoxyethanol, Octoxynol, Polyoxyethylene(10) Octylphenyl Ether, t-Octylphenoxypolyethoxyethanol, O-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]polyethoxyethanol
M-Clarity™ quality level = MQ200
T8787-50ML T8787-100ML T8787-250ML
SIALT8787-50MLEA 49.8 EUR
SIALT8787-50ML SIALT8787-100ML SIALT8787-250ML
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether), Sigma-Aldrich®
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether)
Boiling Pt: ∼200 °C (1013 hPa)
Melting Pt: 6
Density: 1.06 g/cm³ (20 °C)
Flash Pt: 251 °C (closed cup)
Storage Temperature: Refrigerator
MDL Number: MFCD00132505
CAS Number: 9002-93-1
UN: 3082
ADR: 9 III
Merck Index: 13,06793