TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether), Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Polyethyleneglycol-[4-(1,1,3,3-tetramethylbutyl)phenyl]-ether, (p-tert-Octylphenoxy)polyethoxyethanol, Octoxynol, Polyoxyethylene(10) Octylphenyl Ether, t-Octylphenoxypolyethoxyethanol, O-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]polyethoxyethanol
M-Clarity™ quality level = MQ200
X100-1L X100-500ML X100-5ML X100-100ML X100-5GA X100-1GA
SIALX100-1LEA 119 EUR
SIALX100-1L SIALX100-500ML SIALX100-5ML SIALX100-100ML SIALX100-5GA SIALX100-1GA
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether), Sigma-Aldrich®
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether)
Boiling Pt: ∼200 °C (1013 hPa)
Melting Pt: 6
Density: 1.06 g/cm³ (20 °C)
Flash Pt: 251 °C (closed cup)
Storage Temperature: Refrigerator
MDL Number: MFCD00132505
CAS Number: 9002-93-1
UN: 3082
ADR: 9 III
Merck Index: 13,06793