TRITON™ X-114 (Polyethylene glycol tert-octylphenyl ether), Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ200
X114-100ML X114-1L X114-500ML X114-1GA X114-5GA
SIALX114-100MLEA 73.3 EUR
SIALX114-100ML SIALX114-1L SIALX114-500ML SIALX114-1GA SIALX114-5GA
TRITON™ X-114 (Polyethylene glycol tert-octylphenyl ether), Sigma-Aldrich®
TRITON™ X-114 (Polyethylene glycol tert-octylphenyl ether)
Boiling Pt: ∼150 °C (1013 hPa)
Density: 1.055 g/cm³ (20 °C)
Flash Pt: >93 °C
Storage Temperature: Ambient
MDL Number: MFCD00128254
CAS Number: 9036-19-5
UN: 3082
ADR: 9 III