Tween® 80 (Polysorbate), Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
P1754-1GA P1754-500ML P1754-25ML
SIALP1754-1GAEA 375 EUR
SIALP1754-1GA SIALP1754-500ML SIALP1754-25ML
Tween® 80 (Polysorbate), Sigma-Aldrich®
Tween® 80 (Polysorbate)
Boiling Pt: ∼100 °C (1013 hPa)
Melting Pt: –21 °C
Density: 1.062 g/cm³ (20 °C)
Flash Pt: >149 °C
Storage Temperature: Ambient
MDL Number: MFCD00082107
CAS Number: 9005-65-6
EINECS: 500-019-9
Merck Index: 13,07664