Tween® 20 (Polysorbate), Sigma-Aldrich®
Supplier: Merck
Synonyms: Polyoxyethylene sorbitan monolaurate
M-Clarity™ quality level = MQ300
P2287-100ML P2287-500ML
SIALP2287-100MLEA 52.6 EUR
SIALP2287-100ML SIALP2287-500ML
Tween® 20 (Polysorbate), Sigma-Aldrich®
Tween® 20 (Polysorbate)
Formula: C₅₈H₁₁₄O₂₆
MW: 1227.53 g/mol
Density: 1.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00165986
CAS Number: 9005-64-5
EINECS: 500-018-3