Tween® 20 (Polysorbate), Sigma-Aldrich®
Supplier: Merck
Synonyms: Polyoxyethylene sorbitan monolaurate
M-Clarity™ quality level = MQ200
P6585-100ML P6585-10ML
SIALP6585-100MLEA 1750 EUR
SIALP6585-100ML SIALP6585-10ML
Tween® 20 (Polysorbate), Sigma-Aldrich®
Tween® 20 (Polysorbate)
Formula: C₅₈H₁₁₄O₂₆
MW: 1227.53 g/mol
Density: 1.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00165986
CAS Number: 9005-64-5
EINECS: 500-018-3