Polysorbate 20, Sigma-Aldrich®
Supplier: Merck
Synonyms: Polyoxyethylene-20-Sorbitan Monolaurate, Sorbitan monolaurate, ethoxylated
M-Clarity™ quality level = MQ300
W291501-10KG W291501-1KG W291501-25KG W291501-SAMPLE
SIALW291501-10KGEA 266 EUR
SIALW291501-10KG SIALW291501-1KG SIALW291501-25KG SIALW291501-SAMPLE
Polysorbate 20, Sigma-Aldrich®
Polysorbate 20
Formula: C₅₈H₁₁₄O₂₆
Storage Temperature: Ambient
MDL Number: MFCD00165986
CAS Number: 9005-64-5
EINECS: 500-018-3