Tween® 40 (Polysorbate), Sigma-Aldrich®
Supplier: Merck
Synonyms: Polyoxyethylene sorbitan monopalmitate
M-Clarity™ quality level = MQ100
P1504-100ML P1504-500ML P1504-1GA
SIALP1504-100MLEA 52.8 EUR
SIALP1504-100ML SIALP1504-500ML SIALP1504-1GA
Tween® 40 (Polysorbate), Sigma-Aldrich®
Tween® 40 (Polysorbate)
Formula: C₆₂H₁₂₂O₂₆
Boiling Pt: ∼100 °C (1013 hPa)
Melting Pt: ∼0.1 °C
Density: 1.05 g/cm³ (20 °C)
Flash Pt: >149 °C
Storage Temperature: Ambient
MDL Number: MFCD00165345
CAS Number: 9005-66-7