Tween® 85 (Polysorbate), Sigma-Aldrich®
Supplier: Merck
Synonyms: Polyoxyethylenesorbitan Trioleate
M-Clarity™ quality level = MQ200
P4634-100ML P4634-500ML P4634-1GA
SIALP4634-100MLEA 37.3 EUR
SIALP4634-100ML SIALP4634-500ML SIALP4634-1GA
Tween® 85 (Polysorbate), Sigma-Aldrich®
Tween® 85 (Polysorbate)
Density: 1.02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD01779641
CAS Number: 9005-70-3