(±)-2-Bromopropionic acid, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: DL-2-Bromopropanoic acid, 2-Bromopropanoic acid, DL-2-Bromopropionic acid, (±)-2-Bromopropanoic acid

M-Clarity™ quality level = MQ200

B78300-1KG B78300-250G
SIALB78300-1KGEA 180 EUR
SIALB78300-1KG SIALB78300-250G
(±)-2-Bromopropionic acid, Sigma-Aldrich®
(±)-2-Bromopropionic acid
Formula: H₃CBrCHCOOH
Boiling Pt: 203 °C (1013 hPa)
Melting Pt: ∼25 °C
Density: 1.7 g/cm³ (25 °C)
Flash Pt: 100 °C
MDL Number: MFCD00004211
CAS Number: 598-72-1
EINECS: 209-947-6
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR