(±)-2-Chloropropionic acid, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
α-Chloropropionic Acid, (±)-2-Chloropropanoic acid, 2-Chloropropionic acid
M-Clarity™ quality level = MQ100
109274-100G
109274-500G
SIAL109274-100GEA
38.8
EUR
SIAL109274-100G
SIAL109274-500G
(±)-2-Chloropropionic acid, Sigma-Aldrich®
(±)-2-Chloropropionic acid
Formula:
C₃H₅ClO₂ MW: 108.52 g/mol Boiling Pt: 180 Melting Pt: –12 °C Density: 1.27 g/cm³ (20 °C) |
MDL Number:
MFCD00004224 CAS Number: 598-78-7 EINECS: 209-952-3 UN: 2511 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...