(±)-2-Mercaptopropionic acid, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
T31003-100G T31003-500G T31003-5G
SIALT31003-100GEA 54.8 EUR
SIALT31003-100G SIALT31003-500G SIALT31003-5G
(±)-2-Mercaptopropionic acid, Sigma-Aldrich®
(±)-2-Mercaptopropionic acid
Formula: C₃H₆O₂S
Boiling Pt: 205 °C (1013 hPa)
Melting Pt: 10…14 °C
Density: 1.2 g/cm³ (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00004862
CAS Number: 79-42-5
EINECS: 201-206-5
UN: 2936
ADR: 6.1,II
Merck Index: 12,09476