(±)-3-Octanol, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: 3-Octanol, (±)-Ethyl pentyl carbinol, (±)-3-Hydroxy octane, (±)-Pentyl ethyl carbinol, DL-3-Octanol

M-Clarity™ quality level = MQ100

218405-250G 218405-50G
SIAL218405-250GEA 170 EUR
SIAL218405-250G SIAL218405-50G
(±)-3-Octanol, Sigma-Aldrich®
(±)-3-Octanol
Formula: C₈H₁₈O
MW: 130.23 g/mol
Boiling Pt: 174
Melting Pt: –45 °C
Density: 0.819 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004590
CAS Number: 589-98-0
EINECS: 209-667-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR