(±)-3-Octanol, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 3-Octanol, (±)-Ethyl pentyl carbinol, (±)-3-Hydroxy octane, (±)-Pentyl ethyl carbinol, DL-3-Octanol
M-Clarity™ quality level = MQ300
W358118-100G-K W358118-1KG-K
SIALW358118-100G-KEA 108 EUR
SIALW358118-100G-K SIALW358118-1KG-K
(±)-3-Octanol, Sigma-Aldrich®
(±)-3-Octanol
Formula: C₈H₁₈O
Boiling Pt: 174 °C (1013 hPa)
Melting Pt: –45 °C
Density: 0.819 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004590
CAS Number: 589-98-0
EINECS: 209-667-4