(±)-1,5-Hexanediol, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
198188-1G
198188-5G
SIAL198188-1GEA
92.2
EUR
SIAL198188-1G
SIAL198188-5G
(±)-1,5-Hexanediol, Sigma-Aldrich®
(±)-1,5-Hexanediol
Formula:
C₆H₁₄O₂ Boiling Pt: 89 °C (1013 hPa) Density: 0.9809 g/cm³ (0 °C) Storage Temperature: Ambient |
CAS Number:
928-40-5 EINECS: 213-171-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...