3-Mercapto-1,2-propanediol, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: alpha-Thioglycerol, α-Monothioglycerol, Monothioglycerol, (±)-3-Mercapto-1,2-dihydroxy propane, 1-Thioglycerol, (±)-α-Monothioglycerine, (±)-1-Thioglycerine, (±)-3-Mercapto-1,2-propanediol
88640-100ML 88640-500ML
SIAL88640-100MLEA 249 EUR
SIAL88640-100ML SIAL88640-500ML
3-Mercapto-1,2-propanediol, Sigma-Aldrich®
3-Mercapto-1,2-propanediol
Formula: C₃H₈O₂S
Boiling Pt: 118 °C (1013 hPa)
Density: 1.247 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004879
CAS Number: 96-27-5
EINECS: 202-495-0
Merck Index: 12,09471