3-Mercapto-1,2-propanediol ≥97% (by titrimetric analysis), BioReagent, Cell Culture Grade cell culture reagent, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ200
M6145-100ML M6145-250ML M6145-25ML
SIALM6145-100MLEA 253 EUR
SIALM6145-100ML SIALM6145-250ML SIALM6145-25ML
3-Mercapto-1,2-propanediol ≥97% (by titrimetric analysis), BioReagent, Cell Culture Grade cell culture reagent, Sigma-Aldrich®
3-Mercapto-1,2-propanediol
Formula: C₃H₈O₂S
Boiling Pt: 118 °C/5 mmHg (lit.)
Density: 1.25 g/mL (25 °C) (lit.)
Storage Temperature: Refrigerator
MDL Number: MFCD00004879
CAS Number: 96-27-5
EINECS: 202-495-0
Merck Index: 12,09471