(±)-3-Methylvaleric acid, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ300

W343706-100G-K W343706-1KG-K W343706-25G-K
SIALW343706-100G-KEA 212 EUR
SIALW343706-100G-K SIALW343706-1KG-K SIALW343706-25G-K
(±)-3-Methylvaleric acid, Sigma-Aldrich®
(±)-3-Methylvaleric acid
Formula: C₆H₁₂O₂
Boiling Pt: 196 °C (1013 hPa)
Melting Pt: –41 °C
Density: 0.93 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002729
CAS Number: 105-43-1
EINECS: 203-297-7
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR