(±)-2-Methyl-1-butanol, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ300
W399817-100G-K W399817-1KG-K
SIALW399817-100G-KEA 96.3 EUR
SIALW399817-100G-K SIALW399817-1KG-K
(±)-2-Methyl-1-butanol, Sigma-Aldrich®
(±)-2-Methyl-1-butanol
Formula: C₅H₁₂O
MW: 88.15 g/mol
Boiling Pt: 129
Melting Pt: –70 °C
Density: 0.82 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004743
CAS Number: 137-32-6
EINECS: 205-289-9
UN: 1105
ADR: 3 III
Merck Index: 12,06109